siRNA Details
| Gene Symbol: Ssr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025448.3 | NP_079724.1 | 2 | 436 | ||
| GenBank mRNA | AK005340.1 | - | 3 | 307 | ||
| AK010565.1 | - | 3 | 274 | |||
| AK012281.1 | - | 2 | 264 | |||
| AK020145.1 | - | 3 | 257 | |||
| AK050505.1 | - | 2 | 437 | |||
| AK084530.1 | - | 3 | 274 | |||
| BC010214.1 | - | 2 | 420 | 3497894 | ||

Ssr2
Genomic Map