siRNA Details
| Gene Symbol: Ing5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025454.2 | NP_079730.1 | 4, 5 | 437 | ||
| GenBank mRNA | AK006421.1 | - | 4, 5 | 536 | ||
| AK007536.1 | - | 4, 5 | 428 | |||
| AK028010.1 | - | 4, 5 | 415 | |||
| AK028397.1 | - | 4, 5 | 438 | |||
| AK145695.1 | - | 4, 5 | 410 | |||
| BC064674.1 | - | 4, 5 | 436 | 6400183 | ||

Ing5
Genomic Map