siRNA Details
| Gene Symbol: Ccdc28b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025455.2 | NP_079731.2 | 5 | 658 | ||
| GenBank mRNA | AK007427.1 | - | 4 | 435 | ||
| AK009176.1 | - | 3 | 278 | |||
| AK028262.1 | - | 5 | 636 | |||
| AK158016.1 | - | 5 | 662 | |||
| BC028873.1 | - | 5 | 650 | 4220355 | ||

Ccdc28b
Genomic Map