siRNA Details
| Gene Symbol: Fam134b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034851.2 | NP_001030023.1 | 6 | 776 | ||
| NM_001277315.1 | NP_001264244.1 | 4 | 865 | |||
| NM_001277316.1 | NP_001264245.1 | 4 | 448 | |||
| NM_001277317.1 | NP_001264246.1 | 4 | 426 | |||
| NM_001277318.1 | NP_001264247.1 | 4 | 398 | |||
| NM_025459.3 | NP_079735.2 | 5 | 566 | |||
| GenBank mRNA | AK007506.1 | - | 4 | 427 | ||
| AK015182.1 | - | 5 | 388 | |||
| AK146972.1 | - | 4 | 449 | |||
| AK167327.1 | - | 4 | 866 | |||
| BC019494.1 | - | 5 | 505 | 4460992 | ||
| BC054415.1 | - | 4 | 385 | 3487542 | ||

Fam134b
Genomic Map