siRNA Details
| Gene Symbol: Aamdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177945.1 | NP_001171416.1 | 3 | 307 | ||
| NM_001177946.1 | NP_001171417.1 | 3 | 307 | |||
| NM_001177947.1 | NP_001171418.1 | 2 | 203 | |||
| NM_183251.3 | NP_899074.2 | 2 | 203 | |||
| GenBank mRNA | AA065573.1 | - | 2 | 140 | ||
| AK007558.1 | - | 2 | 155 | |||
| AK008195.1 | - | 2 | 148 | |||
| BC026557.1 | - | 3 | 225 | 5027461 | ||
| BC062811.1 | - | 3 | 262 | 1264501 | ||
| BY259812.1 | - | 3 | 304 | |||

Aamdc
Genomic Map