siRNA Details
| Gene Symbol: Smim20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145433.1 | NP_001138905.1 | 2 | 266 | ||
| GenBank mRNA | AK003139.1 | - | 2 | 201 | ||
| AK007474.1 | - | 3 | 237 | |||
| BC030079.1 | - | 2 | 202 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim20| Gene Symbol: Smim20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145433.1 | NP_001138905.1 | 2 | 266 | ||
| GenBank mRNA | AK003139.1 | - | 2 | 201 | ||
| AK007474.1 | - | 3 | 237 | |||
| BC030079.1 | - | 2 | 202 | |||