siRNA Details
| Gene Symbol: Tmem218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025464.2 | NP_079740.1 | 3 | 234 | ||
| GenBank mRNA | AK003010.1 | - | 3 | 231 | ||
| AK007575.1 | - | 3 | 218 | |||
| AK008902.1 | - | 3 | 205 | |||
| AK161115.1 | - | 236 | ||||
| BC034160.1 | - | 3 | 234 | 5049687 | ||

Tmem218
Genomic Map