siRNA Details
| Gene Symbol: Mptx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025470.3 | NP_079746.1 | 2 | 332 | ||
| GenBank mRNA | AK007658.1 | - | 2 | 333 | ||
| AK007963.1 | - | 2 | 332 | |||
| BC024348.1 | - | 2 | 319 | 4985132 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mptx1| Gene Symbol: Mptx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025470.3 | NP_079746.1 | 2 | 332 | ||
| GenBank mRNA | AK007658.1 | - | 2 | 333 | ||
| AK007963.1 | - | 2 | 332 | |||
| BC024348.1 | - | 2 | 319 | 4985132 | ||