siRNA Details
| Gene Symbol: Cenpw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109747.1 | NP_001103217.1 | 2 | 252 | ||
| GenBank mRNA | AK005988.1 | - | 2 | 234 | ||
| AK141272.1 | - | 2 | 234 | |||
| BC050071.1 | - | 204 | ||||
| BC147188.1 | - | 2 | 249 | 9056212 | ||
| BC147189.1 | - | 2 | 249 | 9056237 | ||
| BC147710.1 | - | 2 | 249 | 9087469 | ||
| BC147721.1 | - | 2 | 249 | 9087480 | ||
| BG804382.1 | - | 2 | 252 | |||

Cenpw
Genomic Map