siRNA Details
| Gene Symbol: Senp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK010813.1 | - | 4 | 315 | ||
| AK012740.1 | - | 5 | 407 | |||
| AK013090.1 | - | 5 | 407 | |||
| AK020068.1 | - | 4 | 310 | |||
| AK083086.1 | - | 5 | 391 | |||
| AK085060.1 | - | 5 | 406 | |||
| AK122542.1 | - | 5 | 402 | |||
| AK164354.1 | - | 5 | 391 | |||
| BC058593.1 | - | 5 | 349 | 5364345 | ||
| CF583861.1 | - | 6 | 605 | |||

Senp7
Genomic Map