siRNA Details
| Gene Symbol: Susd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025491.3 | NP_079767.2 | 3 | 327 | ||
| NM_028340.1 | NP_082616.1 | 3 | 496 | |||
| GenBank mRNA | AK003860.1 | - | 3 | 327 | ||
| AK006068.1 | - | 3 | 318 | |||
| AK013276.1 | - | 3 | 319 | |||
| BC045525.1 | - | 3 | 496 | 6468669 | ||

Susd3
Genomic Map