siRNA Details
| Gene Symbol: Susd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025491.3 | NP_079767.2 | 5 | 777 | ||
| NM_028340.1 | NP_082616.1 | 5 | 946 | |||
| GenBank mRNA | AK003860.1 | - | 5 | 777 | ||
| AK006068.1 | - | 5 | 767 | |||
| BC045525.1 | - | 5 | 946 | 6468669 | ||

Susd3
Genomic Map