siRNA Details
| Gene Symbol: Cdrt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025496.1 | NP_079772.1 | 2 | 509 | ||
| GenBank mRNA | AK006125.1 | - | 2 | 509 | ||
| AK006342.1 | - | 2 | 489 | |||
| BC048461.1 | - | 2 | 476 | 6771281 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdrt4| Gene Symbol: Cdrt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025496.1 | NP_079772.1 | 2 | 509 | ||
| GenBank mRNA | AK006125.1 | - | 2 | 509 | ||
| AK006342.1 | - | 2 | 489 | |||
| BC048461.1 | - | 2 | 476 | 6771281 | ||