siRNA Details
| Gene Symbol: Adprm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025510.3 | NP_079786.2 | 3 | 691 | ||
| GenBank mRNA | AK009142.1 | - | 3 | 669 | ||
| AK010520.1 | - | 2 | 653 | |||
| AK145941.1 | - | 3 | 634 | |||
| BC004029.1 | - | 3 | 643 | 3495093 | ||

Adprm
Genomic Map