siRNA Details
| Gene Symbol: Exosc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025513.3 | NP_079789.1 | 3, 4 | 629 | ||
| GenBank mRNA | AK006281.1 | - | 3, 4 | 525 | ||
| AK009165.1 | - | 1, 2 | 41 | |||
| BC023669.1 | - | 3, 4 | 624 | 5012335 | ||
| BC054085.1 | - | 3, 4 | 634 | 3371550 | ||

Exosc3
Genomic Map