siRNA Details
| Gene Symbol: Exosc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025513.3 | NP_079789.1 | 2 | 394 | ||
| GenBank mRNA | AK006281.1 | - | 2 | 290 | ||
| AK012689.1 | - | 2 | 395 | |||
| BC023669.1 | - | 2 | 389 | 5012335 | ||
| BC054085.1 | - | 2 | 399 | 3371550 | ||

Exosc3
Genomic Map