siRNA Details
| Gene Symbol: 2310022A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122767.1 | NP_001116239.1 | 2 | 202 | ||
| NM_175107.5 | NP_780316.3 | 4 | 311 | |||
| GenBank mRNA | AK009941.1 | - | 2 | 202 | ||
| AK077369.1 | - | 2 | 203 | |||
| AK140814.1 | - | 2 | 182 | |||
| BB853451.1 | - | 4 | 312 | |||
| BC023369.1 | - | 4 | 290 | 5038597 | ||

2310022A10Rik
Genomic Map