siRNA Details
| Gene Symbol: 2310022A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122767.1 | NP_001116239.1 | 3 | 285 | ||
| NM_175107.5 | NP_780316.3 | 5 | 394 | |||
| GenBank mRNA | AK009456.1 | - | 3 | 255 | ||
| AK009941.1 | - | 3 | 285 | |||
| AK077369.1 | - | 3 | 286 | |||
| AK140814.1 | - | 3 | 265 | |||
| BB853451.1 | - | 5 | 395 | |||
| BC023369.1 | - | 5 | 373 | 5038597 | ||

2310022A10Rik
Genomic Map