siRNA Details
| Gene Symbol: Chmp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025519.2 | NP_079795.1 | 2 | 264 | ||
| GenBank mRNA | AK009273.1 | - | 2 | 264 | ||
| AK010707.1 | - | 2 | 331 | |||
| AK145033.1 | - | 2 | 268 | |||
| AK160189.1 | - | 2 | 258 | |||
| AK166371.1 | - | 2 | 286 | |||
| BC115983.1 | - | 2 | 254 | 40105850 | ||
| BC117538.1 | - | 2 | 254 | 40105849 | ||

Chmp4c
Genomic Map