siRNA Details
| Gene Symbol: CYP4F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000896.2 | NP_000887.2 | 13 | 2497 | ||
| NM_001199208.1 | NP_001186137.1 | 13 | 2497 | |||
| NM_001199209.1 | NP_001186138.1 | 13 | 2481 | |||
| XM_005259911.4 | XP_005259968.1 | 11 | 2425 | |||
| XM_011528014.2 | XP_011526316.1 | 11 | 2274 | |||
| XM_017026814.1 | XP_016882303.1 | 12 | 2575 | |||
| GenBank mRNA | AB002454.1 | - | 13 | 2497 | ||
| BC136299.1 | - | 13 | 2489 | 9020286 | ||
| DB184202.1 | - | 1 | 76 | |||

CYP4F3
Genomic Map