siRNA Details
| Gene Symbol: Krtap3-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025524.2 | NP_079800.1 | 1 | 564 | ||
| GenBank mRNA | AK009052.1 | - | 1 | 568 | ||
| AK009410.1 | - | 1 | 566 | |||
| AK010142.1 | - | 1 | 565 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap3-3| Gene Symbol: Krtap3-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025524.2 | NP_079800.1 | 1 | 564 | ||
| GenBank mRNA | AK009052.1 | - | 1 | 568 | ||
| AK009410.1 | - | 1 | 566 | |||
| AK010142.1 | - | 1 | 565 | |||