siRNA Details
| Gene Symbol: Ppp1r7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023200.2 | NP_075689.1 | 6 | 569 | ||
| GenBank mRNA | AF222867.1 | - | 6 | 569 | ||
| AK009342.1 | - | 6 | 584 | |||
| AK009870.1 | - | 6 | 562 | |||
| AK031983.1 | - | 1362 | ||||
| AK129024.1 | - | 6 | 586 | |||
| AK145136.1 | - | 6 | 575 | |||
| AK162517.1 | - | 6 | 577 | |||
| BC013524.1 | - | 6 | 570 | 4237290 | ||

Ppp1r7
Genomic Map