siRNA Details
| Gene Symbol: Prl2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025532.3 | NP_079808.1 | 4 | 456 | ||
| GenBank mRNA | AF525155.1 | - | 4 | 407 | ||
| AK005411.1 | - | 4 | 457 | |||
| AK009863.1 | - | 4 | 456 | |||
| BC100329.1 | - | 4 | 446 | 30787440 | ||
| BC111112.1 | - | 4 | 452 | 40050224 | ||
| BC138199.1 | - | 4 | 449 | 8861218 | ||

Prl2b1
Genomic Map