siRNA Details
| Gene Symbol: Ccdc82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025534.2 | NP_079810.2 | 6 | 1266 | ||
| GenBank mRNA | BC040405.1 | - | 4 | 427 | ||
| BC057326.1 | - | 6 | 1266 | 6826080 | ||
| BC098496.1 | - | 5 | 1193 | 30089818 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc82| Gene Symbol: Ccdc82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025534.2 | NP_079810.2 | 6 | 1266 | ||
| GenBank mRNA | BC040405.1 | - | 4 | 427 | ||
| BC057326.1 | - | 6 | 1266 | 6826080 | ||
| BC098496.1 | - | 5 | 1193 | 30089818 | ||