siRNA Details
| Gene Symbol: Sar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025535.2 | NP_079811.1 | 4, 5 | 391 | ||
| GenBank mRNA | AK002327.1 | - | 4, 5 | 368 | ||
| AK010187.1 | - | 4, 5 | 374 | |||
| AK013559.1 | - | 4, 5 | 353 | |||
| AK150906.1 | - | 4, 5 | 347 | |||
| AK155429.1 | - | 4, 5 | 361 | |||
| BC082550.1 | - | 4, 5 | 360 | 30357800 | ||
| BC085178.1 | - | 4, 5 | 344 | 30701201 | ||

Sar1b
Genomic Map