siRNA Details
| Gene Symbol: Commd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025536.2 | NP_079812.1 | 2 | 259 | ||
| GenBank mRNA | AK010045.1 | - | 2 | 260 | ||
| AK013148.1 | - | 1 | 644 | |||
| BC025891.1 | - | 2 | 224 | 5035059 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Commd5| Gene Symbol: Commd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025536.2 | NP_079812.1 | 2 | 259 | ||
| GenBank mRNA | AK010045.1 | - | 2 | 260 | ||
| AK013148.1 | - | 1 | 644 | |||
| BC025891.1 | - | 2 | 224 | 5035059 | ||