siRNA Details
| Gene Symbol: Alkbh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025538.3 | NP_079814.1 | 2 | 383 | ||
| GenBank mRNA | AK007380.1 | - | 2 | 361 | ||
| AK009812.1 | - | 2 | 370 | |||
| BC029677.1 | - | 2 | 350 | 5364485 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alkbh7| Gene Symbol: Alkbh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025538.3 | NP_079814.1 | 2 | 383 | ||
| GenBank mRNA | AK007380.1 | - | 2 | 361 | ||
| AK009812.1 | - | 2 | 370 | |||
| BC029677.1 | - | 2 | 350 | 5364485 | ||