siRNA Details
| Gene Symbol: Psmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025550.2 | NP_079826.2 | 4 | 635 | ||
| GenBank mRNA | AJ252147.1 | - | 4 | 671 | ||
| AK010287.1 | - | 4 | 668 | |||
| BC006869.1 | - | 4 | 635 | 2646325 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmd6| Gene Symbol: Psmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025550.2 | NP_079826.2 | 4 | 635 | ||
| GenBank mRNA | AJ252147.1 | - | 4 | 671 | ||
| AK010287.1 | - | 4 | 668 | |||
| BC006869.1 | - | 4 | 635 | 2646325 | ||