siRNA Details
| Gene Symbol: Psmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025550.2 | NP_079826.2 | 4 | 665 | ||
| GenBank mRNA | AJ252147.1 | - | 4 | 701 | ||
| AK002343.1 | - | 4 | 699 | |||
| AK010287.1 | - | 4 | 698 | |||
| BC006869.1 | - | 4 | 665 | 2646325 | ||

Psmd6
Genomic Map