siRNA Details
| Gene Symbol: Ndufa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023202.4 | NP_075691.1 | 3 | 203 | ||
| GenBank mRNA | AK002476.1 | - | 3 | 184 | ||
| AK003468.1 | - | 3 | 184 | |||
| AK010299.1 | - | 3 | 161 | |||
| BC052817.1 | - | 3 | 167 | 6487401 | ||
| BC055698.1 | - | 3 | 168 | 6411262 | ||
| BX518947.1 | - | 2 | 80 | |||
| KJ650204.1 | - | 3 | 145 | |||

Ndufa7
Genomic Map