siRNA Details
| Gene Symbol: Pcp4l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025557.1 | NP_079833.1 | 3 | 1347 | ||
| GenBank mRNA | AK002772.1 | - | 3 | 1318 | ||
| AK075738.1 | - | 3 | 1317 | |||
| AY304481.1 | - | 3 | 1318 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcp4l1| Gene Symbol: Pcp4l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025557.1 | NP_079833.1 | 3 | 1347 | ||
| GenBank mRNA | AK002772.1 | - | 3 | 1318 | ||
| AK075738.1 | - | 3 | 1317 | |||
| AY304481.1 | - | 3 | 1318 | |||