siRNA Details
| Gene Symbol: Oxld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025560.2 | NP_079836.1 | 2 | 364 | ||
| GenBank mRNA | AK007842.1 | - | 2 | 365 | ||
| AK021150.1 | - | 2 | 352 | |||
| BC147291.1 | - | 2 | 357 | 9056342 | ||
| BC147292.1 | - | 2 | 357 | 9056341 | ||
| BC172027.1 | - | 2 | 327 | 9054313 | ||

Oxld1
Genomic Map