siRNA Details
| Gene Symbol: Uggt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081252.2 | NP_001074721.2 | 22 | 2613 | ||
| GenBank mRNA | AK046834.1 | - | 9 | 1092 | ||
| BC110672.1 | - | 8 | 994 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uggt2| Gene Symbol: Uggt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081252.2 | NP_001074721.2 | 22 | 2613 | ||
| GenBank mRNA | AK046834.1 | - | 9 | 1092 | ||
| BC110672.1 | - | 8 | 994 | |||