siRNA Details
| Gene Symbol: Cdc26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139291.3 | NP_647452.1 | 4 | 474 | ||
| GenBank mRNA | AK004518.1 | - | 1 | 114 | ||
| AK012627.1 | - | 4 | 475 | |||
| AK047055.1 | - | 494 | ||||
| AK086692.1 | - | 953 | ||||
| AK135556.1 | - | 474 | ||||
| BC005775.1 | - | 3 | 401 | 3602527 | ||
| BC114350.2 | - | 2 | 261 | 40053418 | ||

Cdc26
Genomic Map