siRNA Details
| Gene Symbol: Tnfaip8l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025566.3 | NP_079842.2 | 2 | 1064 | ||
| GenBank mRNA | AK004949.1 | - | 2 | 995 | ||
| AK011245.1 | - | 2 | 1026 | |||
| AK090348.1 | - | 2 | 1066 | |||
| AK136962.1 | - | 2 | 1026 | |||
| AK150678.1 | - | 2 | 1055 | |||
| AK153177.1 | - | 2 | 1050 | |||
| BC032199.1 | - | 2 | 993 | 5324284 | ||

Tnfaip8l1
Genomic Map