siRNA Details
| Gene Symbol: 2610528J11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025572.2 | NP_079848.1 | 3 | 422 | ||
| GenBank mRNA | AK009128.1 | - | 3 | 421 | ||
| AK012175.1 | - | 3 | 455 | |||
| AK147115.1 | - | 3 | 420 | |||
| BC022764.1 | - | 3 | 445 | 4925508 | ||

2610528J11Rik
Genomic Map