siRNA Details
| Gene Symbol: 2610528J11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025572.2 | NP_079848.1 | 3 | 866 | ||
| GenBank mRNA | AK009128.1 | - | 3 | 865 | ||
| AK147115.1 | - | 3 | 864 | |||
| BC022764.1 | - | 3 | 889 | 4925508 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610528J11Rik| Gene Symbol: 2610528J11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025572.2 | NP_079848.1 | 3 | 866 | ||
| GenBank mRNA | AK009128.1 | - | 3 | 865 | ||
| AK147115.1 | - | 3 | 864 | |||
| BC022764.1 | - | 3 | 889 | 4925508 | ||