siRNA Details
| Gene Symbol: Nmnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133435.1 | NP_597679.1 | 5 | 857 | ||
| GenBank mRNA | AF260925.1 | - | 5 | 1010 | ||
| AK011137.1 | - | 5 | 879 | |||
| AK012194.1 | - | 5 | 1091 | |||
| AK152407.1 | - | 1345 | ||||
| AY679721.1 | - | 5 | 857 | |||
| BC138500.1 | - | 5 | 925 | 8861520 | ||
| BC138502.1 | - | 5 | 925 | 8861522 | ||

Nmnat1
Genomic Map