siRNA Details
| Gene Symbol: Nmnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133435.1 | NP_597679.1 | 2 | 191 | ||
| GenBank mRNA | AF260925.1 | - | 2 | 344 | ||
| AK011137.1 | - | 2 | 213 | |||
| AK012194.1 | - | 2 | 425 | |||
| AK017631.1 | - | 201 | ||||
| AK152407.1 | - | 251 | ||||
| AY679721.1 | - | 2 | 191 | |||
| BC138500.1 | - | 2 | 259 | 8861520 | ||
| BC138502.1 | - | 2 | 259 | 8861522 | ||

Nmnat1
Genomic Map