siRNA Details
| Gene Symbol: Ptpmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025576.2 | NP_079852.1 | 2 | 479 | ||
| GenBank mRNA | AK003210.1 | - | 1 | 125 | ||
| AK012674.1 | - | 2 | 469 | |||
| AK014691.1 | - | 2 | 473 | |||
| AK151972.1 | - | 2 | 480 | |||
| AK168416.1 | - | 287 | ||||
| BC026750.1 | - | 2 | 458 | 5135841 | ||

Ptpmt1
Genomic Map