siRNA Details
| Gene Symbol: Ska1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164355.1 | NP_001157827.1 | 6 | 704 | ||
| NM_025581.4 | NP_079857.3 | 6 | 768 | |||
| GenBank mRNA | AK006442.1 | - | 6 | 665 | ||
| AK007138.1 | - | 5 | 536 | |||
| AK013233.1 | - | 6 | 666 | |||
| AK021226.1 | - | 6 | 668 | |||
| AK172500.1 | - | 6 | 671 | |||
| BC019940.1 | - | 6 | 619 | 3709649 | ||
| DQ926863.1 | - | 6 | 646 | |||
| DQ926864.1 | - | 6 | 710 | |||

Ska1
Genomic Map