siRNA Details
| Gene Symbol: Fam213b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025582.3 | NP_079858.2 | 3, 4 | 356 | ||
| GenBank mRNA | AK005188.1 | - | 3, 4 | 357 | ||
| AK012999.1 | - | 3, 4 | 355 | |||
| BC030453.1 | - | 3, 4 | 350 | 5401948 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam213b| Gene Symbol: Fam213b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025582.3 | NP_079858.2 | 3, 4 | 356 | ||
| GenBank mRNA | AK005188.1 | - | 3, 4 | 357 | ||
| AK012999.1 | - | 3, 4 | 355 | |||
| BC030453.1 | - | 3, 4 | 350 | 5401948 | ||