siRNA Details
| Gene Symbol: Ctrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025583.2 | NP_079859.2 | 2 | 114 | ||
| GenBank mRNA | AK003060.1 | - | 2 | 123 | ||
| AK003079.1 | - | 2 | 115 | |||
| AK007566.1 | - | 2 | 114 | |||
| AK007765.1 | - | 2 | 114 | |||
| AK007815.1 | - | 2 | 115 | |||
| AK008644.2 | - | 2 | 115 | |||
| AK008729.1 | - | 2 | 115 | |||
| AK008888.1 | - | 2 | 115 | |||
| AK008927.1 | - | 2 | 115 | |||
| AK018554.1 | - | 2 | 115 | |||
| BC061083.1 | - | 2 | 114 | 30316047 | ||

Ctrb1
Genomic Map