siRNA Details
| Gene Symbol: 1700029F12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080777.2 | NP_001074246.1 | 3 | 759 | ||
| NM_001284205.1 | NP_001271134.1 | 3 | 717 | |||
| NM_025585.3 | NP_079861.1 | 3 | 747 | |||
| GenBank mRNA | AK006036.1 | - | 3 | 735 | ||
| AK006488.1 | - | 3 | 715 | |||
| BC049576.1 | - | 3 | 733 | 6772947 | ||
| CF106098.1 | - | 2 | 569 | |||

1700029F12Rik
Genomic Map