siRNA Details
| Gene Symbol: Rpl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025586.3 | NP_079862.1 | 4 | 704 | ||
| GenBank mRNA | AK168807.1 | - | 3 | 696 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpl15| Gene Symbol: Rpl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025586.3 | NP_079862.1 | 4 | 704 | ||
| GenBank mRNA | AK168807.1 | - | 3 | 696 | ||