siRNA Details
| Gene Symbol: Exoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025588.2 | NP_079864.1 | 15 | 1751 | ||
| GenBank mRNA | AK016532.1 | - | 16 | 1878 | ||
| AK050068.1 | - | 15 | 1759 | |||
| BC031207.1 | - | 8 | 793 | |||
| BC048154.1 | - | 15 | 1751 | 6400932 | ||
| BC049102.1 | - | 15 | 1751 | 5702581 | ||

Exoc2
Genomic Map