siRNA Details
| Gene Symbol: Exoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025588.2 | NP_079864.1 | 3 | 260 | ||
| GenBank mRNA | AK016532.1 | - | 4 | 387 | ||
| AK032766.1 | - | 3 | 282 | |||
| AK042744.1 | - | 3 | 307 | |||
| AK050068.1 | - | 3 | 268 | |||
| AK180355.1 | - | 2 | 153 | |||
| BC048154.1 | - | 3 | 260 | 6400932 | ||
| BC049102.1 | - | 3 | 260 | 5702581 | ||

Exoc2
Genomic Map