siRNA Details
| Gene Symbol: Smim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007443.1 | - | 1 | 227 | ||
| AK019298.1 | - | 1 | 202 | |||
| AK036279.1 | - | 190 | ||||
| CX238554.1 | - | 209 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim4| Gene Symbol: Smim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007443.1 | - | 1 | 227 | ||
| AK019298.1 | - | 1 | 202 | |||
| AK036279.1 | - | 190 | ||||
| CX238554.1 | - | 209 | ||||