siRNA Details
| Gene Symbol: Ppdpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025598.2 | NP_079874.1 | 2, 3 | 291 | ||
| GenBank mRNA | AK003282.1 | - | 2, 3 | 292 | ||
| AK012037.1 | - | 2, 3 | 175 | |||
| AK012327.1 | - | 2, 3 | 293 | |||
| AK154758.1 | - | 2, 3 | 332 | |||
| AK184222.1 | - | 1, 2 | 27 | |||
| AK187673.1 | - | 1, 2 | 27 | |||
| BC058516.1 | - | 2, 3 | 329 | 6824261 | ||
| BC091753.1 | - | 2, 3 | 98 | 4218108 | ||
| BC100439.1 | - | 2, 3 | 183 | 6531448 | ||

Ppdpf
Genomic Map