siRNA Details
| Gene Symbol: Slc30a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023214.7 | NP_075703.1 | 9 | 1074 | ||
| GenBank mRNA | AF233322.1 | - | 9 | 936 | ||
| AF529196.1 | - | 9 | 882 | |||
| AK010008.1 | - | 9 | 1074 | |||
| AK075802.1 | - | 9 | 1047 | |||
| BC017136.1 | - | 9 | 1024 | 4911068 | ||
| BC050193.1 | - | 9 | 973 | 30019802 | ||

Slc30a7
Genomic Map